VWR® Basic White SPP Coveralls
Proveedor: VWR
VWR®
414004-338 414004-336 414004-341 414004-340 414004-337 414004-339 414004-335
414004-337CS 1589.65 MXN
414004-337 414004-336 414004-335 414004-341 414004-340 414004-339 414004-338
VWR® Basic White SPP Coveralls
Prendas desechables
Coveralls are manufactured from a spunbonded polypropylene (SPP) fabric that is strong, lightweight, and breathable.
  • Economical, disposable first-line barrier
  • Maximum breathability
  • Low linting
They protect against dirt, grime, and certain dry particulates in nonhazardous environments
Garments feature inset sleeves, front zipper, tapered collar, and elastic cuffs, ankles, and back. No pockets. Serged seams. Also available in blue (30g/m²).
These general-purpose garments are ideal for less critical areas or pre-gowning entry rooms. Coveralls are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.