Heavy Lift Back Belt, OK-1® Safety, OccuNomix
Proveedor: OccuNomix
OK-1® Safety
SS5 XL SS5 XXL SS5 XXXL
39870-108CS 9902.88 MXN
39870-108 39870-106 39870-110
Heavy Lift Back Belt, OK-1® Safety, OccuNomix
Soportes ergonómicos Apoyos Volver
These SS-5 model back belts are premium heavy lifting belts
Designed specifically for heavy lifting, each belt’s rigid inner core includes a built-in lumbar pad that provides support to the lower lumbar region of the spine. The Double Touch fastening system provides firm and comfortable abdominal support. The outer shell utilizes rugged Cordura/Supplex® fabric and 8.9cm (31/2") wide webbing. Width: 13cm (5").