VWR® Fold-Up Magnifiers
3434 3431 3435 3436 3433
62379-547EA 1055.98 MXN
62379-541 62379-545 62379-543 62379-537 62379-547
VWR® Fold-Up Magnifiers
A hands-free magnifier allows two-handed examination of objects.
  • All-glass fabrication of optics
  • Magnifiers fold up for easy, compact storage
  • White gridded markings for measurement
  • Battery-powered illumination
The all-glass optics assure perfect focus with zero distortion. Provide a sharp, crisp, distinct image with vivid enhancement of details. Two sizes (62379-537 and -547) feature battery-powered brilliant illumination for precision work (batteries supplied). Bottom panel features white gridded markings in both inches and millimeters for easy measurement. Magnifiers fold twice for compact storage. The lighted hands-free magnifier 62379-547 with 10X magnification is elongated and does not fold.