KONTES® Graham Condensers, Kimble Chase
Proveedor: DWK Life Sciences
KONTES®
439000-2420 439000-2430 439000-2440 439000-2425
KT439000-2420EA 2909.89 MXN
KT439000-2430 KT439000-2440 KT439000-2420 KT439000-2425
KONTES® Graham Condensers, Kimble Chase
Condensadores
Vapor travels through a coil extending through the length of the condenser and is surrounded by a cooling jacket
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Standard Taper outer joint at top and standard taper inner drip joint at the bottom.