Sharps Disposal Containers, Covidien
Proveedor: Covidien
8950SA 8527R 8970 8920SA 8990SA
15704-196CS 8879.27 MXN
15704-196 95042-982 15704-210 15704-203 15704-192
Sharps Disposal Containers, Covidien
Recipientes Contenedores de objetos punzantes
Ideal for lancets, scalpels, slides, pipets, and syringes
Rotor lids feature a locking rotor to secure lid. Hinged lids permit temporary closure when not in use. All containers lock for final disposal. Container 15704-214 features a hinged, sealing gasket lid to provide leak-resistant containment of contents. Adhesive-backed red plastic holder fits disposal container 15704-196.
Certificaciones: Containers meet or exceed JCAHO, OSHA, and NIOSH/CDC standards and requirements.