PYREX® Graham Condensers, Corning
Proveedor: Corning
PYREX®
2560-300 2560-500 2560-200 2560-400
89090-724CS 5383.33 MXN
89091-370 89090-724 23139-041 89091-434
PYREX® Graham Condensers, Corning
Condensadores
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These Graham condensers have a high surface area per unit with a standard taper outer joint at the top and a standard taper inner joint at the bottom with a drip tip. They are primarily used in vacuum distillations. Condenser tubulations are approximately 10 mm ext.Ø.