Orion™ AQ4500 Turbidimeter, Thermo Scientific
Proveedor: Thermo Scientific
ORION®
AQ4500 AQ4CBL AQ4500
14233-240EA 37209.83 MXN
14217-922 14233-240 CA14233-240
Orion™ AQ4500 Turbidimeter, Thermo Scientific
Turbidímetros
Utilize dual LED light sources and drinking water/wastewater methods approved for U.S.
  • Log data into internal memory, or use the optional RS232 cable to send data to printer or download to a computer for easy time and date stamped results storage
  • Nephelometric and ratiometric measurements with autoranging
  • IP 67 waterproof portable turbidity meter
  • Obtain readings in the range of 0-4000 NTU with a choice of units: NT, FTU, FNU, ASBC, or EBC
The Orion™ AQ4500 Turbidity Meter is equipped with advanced features, including dual light sources and drinking water/wastewater methods approved for U.S. EPA regulatory reporting. Keep pace with changing regulatory protocols, use the White Light LED to comply with EPA 180.1 requirements or Infrared (IR) LED to comply with ISO 7027 requirements. Select from measure modes including EPA 180.1, ISO Nephelometric, ISO Absorbance, % transmittance, EBC, and ASBC. This IP67 waterproof meter records 100 data points and offers an average battery life of 2500 hours intermittent use.
EPA regulatory reporting offered by the Orion™ AQ4500 Turbidity Meter.
Certificaciones: US EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg 38351-38356.EPA 180.1 and ISO 7032.
Información para pedidos: Includes AQ4500 meter, Set of 5 USEPA approved primary calibration standards, seven glass vials, user guide, and carrying case. Two-year limited warranty included. Reorder replacement Calibration Standards for AQ4500 (AC45ST) and replacement vials in a package of 12 (AC2T24). Silicone oil and cloth set (AC45S1) is also available separately.