VWR® Maximum Non-Skid High-Top Boot Covers
Proveedor: VWR
VWR®
414004-417 414004-419 414004-418 414004-415 414004-414 414004-416
414004-414CS 5412.33 MXN
414004-414 414004-419 414004-418 414004-417 414004-416 414004-415
VWR® Maximum Non-Skid High-Top Boot Covers
Prendas desechables
These non-skid boot covers are engineered to optimize barrier protection, cleanliness, comfort, and skid-resistance properties
  • Superior coefficient of friction ensures maximum traction
  • Maximum skid resistance on wet and smooth surfaces
  • Serged seams for extra protection
  • Clean, low-linting fabric
Boot covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
A high vapor transmission rate keeps the wearer comfortable, even throughout extended use. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.