VWR® Signature™ Disposable Electroporation Cuvettes
Proveedor: VWR
VWR® Signature™
4040-04 1040-04S 4010-01 1010-01 1040-04 4020-02 1020-02
89047-208CS 249704.07 MXN
89047-208 89047-206 76102-580 76102-576 89047-210 76407-204 76102-578
VWR® Signature™ Disposable Electroporation Cuvettes
Cubetas Cubetas de electroporación
VWR Signature™ Disposable Electroporation Cuvettes are designed for use with bacterial, yeast, fungi, and mammalian cells.
  • Cuvettes are precision built to provide exacting tolerances, ensuring reproducible results
  • Polished aluminum plates reduce static and arcing
  • Compatible with most electroporation devices
  • Gamma irradiated for sterility
Disposable cuvettes are manufactured from high quality, durable polycarbonate material that has been tested to optimize transformation efficiencies.
Información para pedidos: Each pack contains 50 individually wrapped cuvettes.