VWR® Spectrosil Spectrophotometer Cells
Proveedor: VWR
VWR®
14222-706 14222-704 14222-708 58016-618 58016-254 414004-229 58016-469 58016-210 58016-378 58016-695 58016-276 14231-068 58016-704 58016-505 14222-710 14222-712 14222-702 58016-480 58016-699 14222-700 58016-673 58016-298
414004-069CS 141913.83 MXN
414004-049 414004-059 414004-069 414004-068 414004-078 414004-055 414004-229 414004-065 414004-054 414004-076 414004-053 414004-052 414004-074 414004-063 414004-051 414004-062 414004-073 414004-061 414004-050 414004-071 414004-060 414004-070
VWR® Spectrosil Spectrophotometer Cells
Cubetas Cubetas para espectrofotómetros
Manufactured from Spectrosil quartz for accurate readings over the entire UV range to near infrared range.
  • Far UV quartz
  • Wavelength range of 170–2700 nm
  • Matched sets available
  • Fully heat-fused construction
Due to the fully heat-fused construction (no intermediate materials or adhesives) and careful annealing to remove any strain, there is no optical or physical distortion of the two optically polished windows
In semi-micro cells, the walls parallel to the light path are thickened, decreasing the sample chamber width to 4mm. Sub-micro cells have a Z dimension that must be matched between the cell and the instrument to be used.
Three cover types are available: flat lid, stopper, and screw cap. Flat lids and stoppers are made of polytetrafluoroethylene (PTFE). Stoppered cells feature a block fused to the top of the cell with a ground hole for the stopper, providing a more suitable seal for volatile liquids. Cells with screw caps can be used under anaerobic conditions and offer an air-tight fit. Cell 414004-054 features a closed-top polymer screw cap with a silicone rubber gasket. Cell 414004-055 features an open-top polymer screw cap with a silicone rubber septum.
Note: Not recommended for use with ultrasonic cleaning.
Información para pedidos: For Spectrosil cells for use in fluorometers, see 414004-064 series. Packs of two cells are matched sets.