VWR® Optical Glass Spectrophotometer Cells
Proveedor: VWR
VWR®
58016-425 14235-606 58016-697 58016-414
414004-067CS 40229.11 MXN
414004-067 414004-077 414004-066 414004-075
VWR® Optical Glass Spectrophotometer Cells
Cubetas Cubetas para espectrofotómetros
Obtain accurate, dependable results with these economical alternatives for spectrophotometric analyses in the visible and near infrared regions.
  • Matched sets
  • Fully heat-fused construction
Due to the fully heat-fused construction (no intermediate materials or adhesives) and careful annealing to remove any strain, there is no optical or physical distortion of the two optical windows. Matched cells fall within the accepted pathlength standard of ±0.01mm. Cells exhibit a transmission tolerance within 0.5% at 365nm. Lids and stoppers are made of polytetrafluoroethylene (PTFE).
Información para pedidos: Items 414004-066 and 414004-067 are sold in matched sets of two and four cells, respectively.