Acurex™ 501 and 502 Compact Bottle-Top Dispensers, DWK Life Sciences
Proveedor: DWK Life Sciences
Acurex™
W844055 844064 W844057 844068 844060 W844058 W844056 W844059
89044-740EA 17684.18 MXN
53501-632 89044-740 53501-634 14231-604 89044-736 89044-738 89044-732 89044-734
Acurex™ 501 and 502 Compact Bottle-Top Dispensers, DWK Life Sciences
Dispensadores de la parte superior del frasco
Compact dispensers for safe reagent handling, adapted to refrigerator storage as well as water bath heating
Dosing mechanism entirely protected inside reservoir. Manufactured and tested to fully comply with the latest regulations on instrument safety and precision. Volumes range from 0.2 to 30 mL. This product is also fully autoclavable at 121°C (250°F).
Información para pedidos: One-year manufacturer's warranty.