Straight-Sided Jars, Amber Glass, Kimble Chase, DWK Life Sciences
Proveedor: DWK Life Sciences
5420458V-21 5420870C-26 5420870V-21 5421689C-26 5420458C-26 5420458V-26
89046-010CS 2408.96 MXN
89046-010 89046-012 89045-994 89045-898 89046-006 89045-902
Straight-Sided Jars, Amber Glass, Kimble Chase, DWK Life Sciences
Tarros Tarros para uso general
Jars feature a wide-mouth design for efficient addition and removal contents
Amber-colored glass protects light-sensitive contents. Made of Type III soda-lime glass. Available with or without black phenolic closures.
Información para pedidos: Packaging options include bulk packs (shrink-wrapped modules with caps in bags), convenience packs (caps attached), or pallet packs (jars only; caps not included). All 1L (33.8oz.) jars are packed in corrugated cartons with divider cells. Pallet packs of 1L (33.8oz.) jars contain 54 cartons.