VWR® Disposable Aluminum Weighing Dishes
Proveedor: VWR
VWR®
D200-50 D500-50 D44-100 D75-100 D63-100
25433-018PK 487.36 MXN
25433-018 25433-016 25433-020 25433-014 25433-022
VWR® Disposable Aluminum Weighing Dishes
Navecillas Navecillas de pesaje
Ideal for weighing, sampling, dispensing, storage, food processing, evaporating or general laboratory use.
  • Made of light-gauge aluminum
  • Stackable
  • Contaminant-free
  • Petroleum-free
Pans feature a flanged edge with curled lip and are contaminate- and petroleum-free.