VWR® Disposable Aluminum Weighing and Drying Pans
Proveedor: VWR
VWR®
D-126-50 D-11 D-12 D-125 D-123
12177-305CS 4284.13 MXN
12177-305 12177-266 12177-299 12177-301 12175-001
VWR® Disposable Aluminum Weighing and Drying Pans
Navecillas Navecillas de pesaje
These oil-free pans are ideal for general-purpose weighing, dispensing, storing, and drying applications.
  • Sturdy construction
  • Compatible with most balances and dryers
  • Ideal for general-purpose applications
  • Disposable
  • Aluminum construction
Sturdy, with a slight taper for easy stacking. The 7 cm (23/4") I.D. pan is ideal for enclosed balances and weighing equipment with a 7 to 9.5 cm (23/4 to 33/4") diameter head.