VWR® AlcoSCRUB® Instant Antiseptic Hand Sanitizer and Cleanser
Proveedor: VWR
VWR® AlcoSCRUB®
42016-12-601640 42032-12-601 42016-12-601640 42032-12-601640
56700-116CS 3855.9 MXN
CA56700-116 CA56700-148L 56700-116 56700-148
VWR® AlcoSCRUB® Instant Antiseptic Hand Sanitizer and Cleanser
Limpieza de la piel Desinfectantes de la piel Desinfectantes para la piel líquidos y en espuma
Fast-acting antimicrobial gel is designed for use when water is not available to supplement hand washing
  • Fast-acting antimicrobial gel for use when water is not available
  • Ideal for reducing bacteria count on hands prior to donning gloves and entering aseptic environment
  • Dries quickly and leaves no sticky residue
  • Emollients help moisturize hands and keep them chap-free
  • Contains 62% ethyl alcohol formula
Gel dries quickly and leaves no sticky residue. Emollients help moisturize hands and keep them chap-free. Contains 62% ethyl alcohol and is highly effective against a broad spectrum of microorganisms.
Ideal for reducing bacteria counts on hands prior to donning gloves and entering an aseptic environment.
Información para pedidos: A wall-mounted dispenser (56700-138) for the 800mL (27oz.) Bag-in-a-Box is available; contact your Avantor representative for more information.