Apparel Storage Bins/Dispensers, S-Curve Technologies
Proveedor: S-Curve Technologies
SBD-17
32948-844EA 6835.14 MXN
32948-844
Apparel Storage Bins/Dispensers, S-Curve Technologies
Dispensadores de EPI
Designed for storage and dispensing of bouffants, shoe covers, beard covers, and bulk gloves
The hinged lid allows easy access. Constructed of clear, sturdy, 6 mm (¹/₄") thick acrylic or PETG. Mounts on wall for quick, convenient dispensing. Mounting bracket included.