VWR® 4-Place Pipette Racks
Proveedor: VWR
VWR®
HS20611V
82024-536EA 3520.19 MXN
82024-536
VWR® 4-Place Pipette Racks
Soportes de pipetas
These four-place acrylic pipette racks keep instruments secure and conveniently accessible in the laboratory environment
Racks accommodate 0.1 to 25 mL pipettes or cans. Clear plastic construction allows for easy viewing of enclosed pipettes. Racks feature four angled compartments.