Nunc™ Biobanking and Cell Culture Cryogenic Tubes, Thermo Scientific
Proveedor: Thermo Scientific
Nunc® CryoTube®
366656 379189 347643 379189 379146 375353 337516 351934 375353 363401 367997 368632 347597 373420 367997 377224 373530 375418 366524 347627 347643 363452 377267 351934 337516 368632 366524 373530 379146 347627 377267 366656 363452 377224 363401 347597 375418
66021-987CS 46772.27 MXN
CA66021-992 CA66021-989 55710-280 66021-989 66021-991 CA66021-994 66021-984 73521-388 66021-994 66021-985 CA73521-390 66021-987 55710-282 89233-142 73521-390 CA66021-993 73521-444 CA66021-996 66021-988 55710-284 CA55710-280 66021-990 66021-986 CA73521-388 CA66021-984 CA66021-987 CA66021-988 CA73521-444 CA66021-991 CA55710-284 CA66021-986A 66021-992 CA66021-990A 66021-993 CA66021-985 CA55710-282 66021-996
Nunc™ Biobanking and Cell Culture Cryogenic Tubes, Thermo Scientific
Viales Criotubos
These cryogenic storage tubes and accessories are designed to work together and provide the best solutions for the most trusted sample storage.
  • Low cellular adhesion and low protein binding
  • Non-cytotoxic, non-mutagenic plastics and pyrogen-free, IVD compatible vials ensure sample integrity
  • Internally-threaded tubes suitable for storage down to the vapor phase of liquid nitrogen
  • Externally threaded options can store at higher general refrigeration temperatures (4 °C)
Applications includes: Regulated applications, irreplaceable high value content storage (down to VPLN), tissue, cells, blood storage for biobanking.
Certificaciones: Compliance with IATA for shipping and CE marked for diagnostic sample storage in the EU. Certification of SAL 10⁻⁶ for the highest sterility assurance available.