VWR® Talon® PVC Coated Open Extension Rings
Proveedor: VWR
VWR® Talon®
80863386 80863385 80863384
97019-056EA 2161.88 MXN
97019-056 97019-054 97019-058
VWR® Talon® PVC Coated Open Extension Rings
Anillos de retorta
Ideal for supporting funnels, round-bottom flasks, reaction vessels, and other apparatuses requiring lower support.
  • PVC coating
  • Fully adjustable depth
  • Capable of supporting a wide range of glassware
These PVC-coated aluminum rings feature an opening that allows simple removal of sample containers. The long extension arm permits depth adjustment of the open ring from the lab frame or ring stand. PVC coating protects glassware.