Nalgene® SYSTEM 100™ Cryogenic Vials, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene® System 100™
5000-1012 5000-1020
66008-710CS 8646.53 MXN
66008-708 66008-710
Nalgene® SYSTEM 100™ Cryogenic Vials, Thermo Scientific
Viales
These vials are designed to increase storage capacity from 81 to 100 vials in mechanical and vapor-phase (LN₂) freezers when used with SYSTEM 100 CryoBox™ vial boxes.
  • Externally threaded vials for aseptic technique
  • White marking area, graduations, and fill line printed on vials
  • Radiation sterilized
  • Non-cytotoxic and non-pyrogenic
  • Self-standing
They are constructed of polypropylene and can withstand temperatures from –196 to 121 °C (–321 to 250 °F). Silicone gasketed polypropylene closures ensure leakproof performance in a microcentrifuge (up to 8000 g) and during shipment.
Certificaciones: CE marked.
Envase: Packaged 25 vials per bag, 20 bags per case.