VWR® Vials, Borosilicate Glass, with Phenolic Screw Cap
Proveedor: VWR
VWR®
035/12-PHC73 029/57-PHC63 022/18-PHC53 014/79-PHC73 005/55-PHC23 040/66-PHC73 001/85-PHC13 003/70-PHC23 014/79-PHC43 007/39-PHC33 011/10-PHC33 022/18-PHC73
66012-088CS 4967.15 MXN
66012-066 66012-022 66012-088 66012-044 66011-100 66011-165 66011-143 66011-121 66011-020 66011-063 66011-041 66011-085
VWR® Vials, Borosilicate Glass, with Phenolic Screw Cap
Viales
These general-purpose vials are available in either a short form or capsule style.
  • Borosilicate glass fabrication
  • Short form or capsule style
  • Cap is flush to vial shoulder
  • Black phenolic molded screw cap
  • Polyvinyl-faced pulp liner
  • General-purpose use
Vials have black phenolic molded screw cap with polyvinyl-faced pulp liner. The cap on the vial is the same diameter as the body, fitting flush to the shoulder.