Scintillation Vials, Borosilicate Glass, with Screw Cap, Wheaton
Proveedor: DWK Life Sciences
986580 986586 986541 986582 986540 986568 986560 986561 986542 986581 986548 986562 986546 986590 986591
66021-453CS 11994.38 MXN
66021-453 66021-497 66021-675 66021-577 66021-533 66021-452 56941-386 66021-600 66021-645 66021-668 66021-657 66021-624 66021-646 66021-602 56941-513
Scintillation Vials, Borosilicate Glass, with Screw Cap, Wheaton
Viales
Made of low-potassium borosilicate glass to critical manufacturing tolerances, ensuring reliable, consistent results in scintillation counting.
  • Background counts remain low and ultraviolet transmission high
  • Vials are designed to load properly into scintillation counting equipment without jamming or breaking
Tight-fitting caps have tops suitable for writing. Vials with attached or unattached screw-caps.
Información para pedidos: Available with a variety of caps and liners. Packaged in individual cells in trays of 100.