D/RNAse Free™ Decontaminant, Argos Technologies
24411-88
47751-044CS 14651.54 MXN
47751-044
D/RNAse Free™ Decontaminant, Argos Technologies
Surface decontaminant specially formulated to eliminate RNase and DNase from laboratory glassware, plastics, stainless steel, and other surfaces
Ideal for cleaning equipment, pipettors, gel boxes, and labware that cannot be autoclaved. Nontoxic formula replaces carcinogenic chemicals such as DEPC. Product is non-biohazardous, nonabrasive, and leaves no residue that can interfere with gel polymerization or enzymatic reactions.