SP Bel-Art Aquet® Detergent for Glassware and Plastics, Bel-Art Products, a part of SP
Proveedor: Bel-Art Products, a part of SP
Aquet®
F17094-0020 F17094-0050 F17094-0030
75783-970EA 1506.46 MXN
75783-970 74950-097 54848-004
SP Bel-Art Aquet® Detergent for Glassware and Plastics, Bel-Art Products, a part of SP
Soluciones de limpieza Detergentes y Limpiadores de Superficie
Safe for glassware, plastics, and surfaces, this phosphate-free, nonionic surfactant will not etch or mar precision glassware.
  • Convenient, pre-portioned pouches make one gallon of 1% solution
  • 3.8 L (1 gal.) bottle comes with 473 mL (16 oz.) working solution dispenser bottle
  • 20 L (5 gal.) Cubitainer™ is available for frequent users
  • Not for use in automatic dishwashers
It rinses quickly and dries without wiping.