Socorex™ Calibrex™ 520 Bottle-Top Dispensers, DWK Life Sciences
Proveedor: DWK Life Sciences
Socorex™ Calibrex™
844002 844000 844004
53501-140EA 16618.54 MXN
53501-140 53501-142 53501-144
Socorex™ Calibrex™ 520 Bottle-Top Dispensers, DWK Life Sciences
Dispensadores de la parte superior del frasco
These chemically inert bottle-top dispensers feature a series of permanently calibrated stops that provide quick and accurate volume selection
Calibration is easily adjusted in the laboratory for temperature or viscosity changes. The volume display is adjustable to four positions for optimal user viewing and ease of setting. The airflow orifice can be modified to accept a membrane filter that eliminates aerosol contamination. The syringe barrel is encased in a protective sleeve, preventing breakage and eliminating spills. The dispenser is autoclavable when fully assembled and easily disassembled for cleaning. Without adapters, dispensers fit a 33mm screw thread bottle.
Reproducibility: <±0.1%
Información para pedidos: Supplied with adapters to fit bottles with 25, 28, 38, and 45mm screw neck. Reservoir bottle not included.