VWR® Aluminum Support Jacks
Proveedor: VWR
VWR®
80863326 80863328 80863322 80863324
12620-902EA 4516.37 MXN
12620-902 12620-904 10027-578 12620-900
VWR® Aluminum Support Jacks
Elevadores de laboratorios
Aluminum lab jacks provide stable height adjustment for various items in the labs such as flasks, baths, and small equipment.
  • Exceptional stability and durability
  • Aluminum construction
  • Four convenient sizes
Oversized, sure-grip adjustment knobs provide smooth and accurate height adjustment
Support rod kit includes a 43.2 cm (17") vertical support rod for mounting to the upper deck of a 15.2x15.2 cm (6x6") or 20.3x20.3 cm (8x8") support jack. Ideal for creating an adjustable support stand for mounting various items such as thermometer clamps, temperature probes, or flask and column clamps. Kit also includes a jam nut and a flat washer.
Top and bottom decks are constructed of anodized aluminum. Internal supports and drive screw are constructed of stainless steel. Lab jacks are designed to hold the specified weight after it is lifted to the preferred height.