VWR® 8" Wash Sieves
Proveedor: VWR
VWR®
140BS8W4 100BS8W4 400BS8W8W/SU 450BS8W8W/SU 400BS8W4 230BS8W4 200BS8W8W/SU 200BS8W4W/SU 230BS8W8 270BS8W4W/SU 270BS8W8W/SU 500BS8W8W/SU 200BS8W8 635BS8W4W/SU 325BS8W4W/SU 325BS8W8 230BS8W4W/SU 170BS8W8 120BS8W8 100BS8W8 270BS8W8 140BS8W8 120BS8W8W/SU 500BS8W4 100BS8W8W/SU 325BS8W4 170BS8W4W/SU 500BS8W4W/SU 270BS8W4 635BS8W8 140BS8W8W/SU 400BS8W4W/SU 170BS8W4 100BS8W4W/SU 140BS8W4W/SU 200BS8W4
15555-700EA 5521.39 MXN
15555-740 15555-782 15555-760 15555-700 15555-766 15555-722 15555-786 15555-720 15555-764 15555-742 15555-726 15555-748 15555-768 15555-746 15555-724 15555-702 15555-718 15555-716 15555-790 15555-772 15555-750 15555-792 15555-732 15555-776 15555-754 15555-710 15555-774 15555-730 15555-752 15555-714 15555-758 15555-756 15555-734 15555-712 15555-708 15555-728
VWR® 8" Wash Sieves
Tamices Tamices de ensayo
VWR® 8" Diameter (203.2 mm ) brass framed wash sieves with stainless steel wire cloth are designed for hand washing of fine particles in pellet, ground, granular or powdered form samples.
  • Designed for use with a variety of sample types
  • Used in a number of industries
These sieves are used in various industries, such as civil engineering, ceramics, paint/pigment and many other applications. Wash sieves are available with #20 stainless steel back-up support cloth to help extend sieve life.
Información para pedidos: Sieves are available in 4" or 8" depths. Sieve depth is measured from the fram rim to the woven wire cloth. Sieve brushes are available as accessories.