Vesphene® IIse Non Sterile Disinfectant Cleaner, STERIS®
Proveedor: Steris
Vesphene®
646108
21899-870CS 8154.3 MXN
21899-870
Vesphene® IIse Non Sterile Disinfectant Cleaner, STERIS®
Soluciones de limpieza Desinfectantes
This one-step, alkaline, phenolic, concentrate cleaner and detergent is specifically formulated for use on hard, non porous surfaces
  • Phenolic active ingredients provide broad spectrum antimicrobial efficacy.
  • Formulation includes surfactants for surface cleaning while disinfecting.
  • Ideal for use in a contamination control rotation.
Vesphene® IIse has a use dilution pH of 10.4 to 10.6 and is ideal for use in a disinfectant rotation program with LpH® se nonsterile disinfectant cleaner (see 16200-146).