KONTES® RNase-Free PELLET PESTLE® Grinders, Kimble Chase
Proveedor: DWK Life Sciences
KONTES® PELLET PESTLE®
749521-1590 749521-0590 749510-1590 749520-0590 749510-0590 749520-0090
KT-749521-1590CS 4059.38 MXN
KT749520-0090 KT749521-0590 KT749510-1590 KT749510-0590 KT-749521-1590 KT749520-0590
KONTES® RNase-Free PELLET PESTLE® Grinders, Kimble Chase
Homogenizadores Trituradoras tisulares
Ideal for grinding soft tissues and resuspension of protein and DNA pellets
Disposable pestles and microtubes are RNase-, DNase-, and pyrogen-free. Individually wrapped to ensure purity. Specifically designed for use with their matching microtubes, unique pestles eliminate the need for pretreatment of pestles and microtubes. Pestle may be hand-operated, or driven by optional cordless mixer motor. Pestle length: 70mm.
Información para pedidos: Mixer motor includes two AA size batteries and adapter for attaching pestle.