KONTES® PELLET PESTLE® Grinders, Kimble Chase
Proveedor: Kimble Chase
KONTES® PELLET PESTLE®
749510-1500 749515-1500 749515-5000 749515-0000 749530-5000 749560-1500 749521-0500 749516-0500 749520-0500 749560-0500 749510-1501 749530-1500 749521-1500 749520-0000
KT749520-0000PK 2094.51 MXN
KT749515-1500 KT749530-5000 KT749515-5000 KT749560-0500 KT749560-1500 KT749521-0500 KT749521-1500 KT749520-0000 KT749515-0000 KT749510-1500 KT749510-1501 KT749520-0500 KT749516-0500 KT749530-1500
KONTES® PELLET PESTLE® Grinders, Kimble Chase
Morteros y manos de mortero Manos de mortero
Grinders are ideal for resuspending protein and DNA pellets or grinding soft tissues
Disposable, autoclavable polypropylene pestles eliminate the need for pretreatment of pestles and microtubes. The CTFE/stainless steel pestles are reusable.
Pestles are specially designed to fit either 0.5mL or 1.5mL microtubes. All pestles and microtubes are autoclavable.
Información para pedidos: Pestle may be hand-operated, or driven by optional cordless mixer motor. Mixer motor includes two AA size batteries and adapter for attaching pestle.