VWR® Talon® S-Connectors
Proveedor: VWR
VWR® Talon®
80862712 80862639
60097-056CS 7958.41 MXN
12621-246 60097-056
VWR® Talon® S-Connectors
Abrazaderas Adaptadores en cruz
Clamp is ideal for constructing lab frames or other supports requiring the connection of two perpendicular rods.
  • electropolished stainless steel or nickel-plated zinc
  • Perpendicular support rods
  • Separate adjustment screws for each rod
Clamp connects two 13mm (1/2") rods at a 90° angle and features separate adjustment screws for each rod location.