VWR® Talon® Lab Frame Feet
Proveedor: VWR
VWR® Talon®
80862644 80862713
60079-257CS 19438.65 MXN
12621-248 60079-257
VWR® Talon® Lab Frame Feet
Varillas de retorta
Strong, durable lab frame foot is designed for mounting lab frames permanently to bench tops, fume hoods, walls, and floors.
  • Electro-finished stainless steel or nickel-plated zinc construction
  • Strong and durable
  • Designed for mounting lab frames permanently
  • Allows easy clean-ups
Accepts 13 mm (1/2") rods. Spilled fluids roll right off the smooth body contour, minimizing contamination and allowing easy clean-ups. Features a set screw and includes three 19 mm (3/4") mounting screws. Stainless steel lab frame foot has a matte finish, and includes stainless steel hardware.