Micro-Mate® Hypodermic Interchangeable Glass Syringes, Cadence Science®
Proveedor: Cadence Science
Micro-Mate®
5037 5038 5036 5007 5067 5016 5024 5006 5004 5005 5034 5027 5015 5049 5014 5058 5048
20068-512EA 952.54 MXN
20068-060 20068-512 20067-990 20067-982 20068-450 20068-518 20068-059 20068-524 20068-054 20068-522 89234-138 20068-080 20068-520 20068-504 20068-056 20068-502 20068-506
Micro-Mate® Hypodermic Interchangeable Glass Syringes, Cadence Science®
Jeringas Jeringas médicas
Interchangeable syringes are manufactured to ensure the perfect mating of barrels and plungers
Precision shrinking ensures improved fit. Colored plunger ends are fused around bottom beveled rim of plunger to facilitate dosage measurements. Made from borosilicate glass providing protection against thermal shock. Corrosion resistant. Graduations permanently fused into glass barrel. Reinforced flanges prevent syringe rolling.
Any Micro-Mate® interchangeable syringe of a particular size will fit any Micro-Mate® interchangeable barrel of that same size