Stereo Microscopes, K Series, Motic
Proveedor: Motic
1101001701642 1101001400992 1100200900211 1100200900281 1101001400641 1101001700752 1100200900261 1100200900322 1101001701642 1100200900311 1101001700672 1101001700672 1101001701642 1101001700752 1101001700712 1101005900092 1101001400852 1101001700801 1101001700712 1100200900291 1101001700801 1101005900082
19000-210EA 49245.3 MXN
CA19000-250 CA19000-272 CA19000-284 CA19000-270 CA19000-282 19000-274 CA19000-276 CA19000-254 CA19000-298 19000-210 CA19000-222 CA19000-288 CA19000-274 CA19000-296 CA19000-286 CA19000-280 CA19000-204 CA19000-278 CA19000-268 CA19000-224 CA19000-218 CA19000-216
Stereo Microscopes, K Series, Motic
Microscopios
These stereo microscopes feature a pair of 10x, 23mm super widefield eyepieces and a 45° inclined binocular tube with dioptric compensation on both tubes and interpupillary distance adjustment between 54mm and 76mm
Choose from the K-400 series microscopes with a four-step magnification changer at 6x, 12x, 25x, and 50x ratios; the K-500 series microsopes with a five-step magnification changer at 6.4x, 10x, 16x, 25x, and 40x ratios; or the K-700 series microscopes that feature a zoom range at 1:5.2 ratio designed on self-centering dovetail ring mechanism, total parfocality, and absolute centering.
Microscopes come in either a streamlined stand with a large working surface or a light stand with built-in incident and transmitted 12V, 10W halogen light with intensity control and enhancing choices of top, bottom, or top and bottom light. Microscopes with the built-in halogen light feature dual voltage with a 110V main supply or 220V.
All microscopes come with a standard 1.0x objective.
Información para pedidos: Heads with focusing mounts, eyepieces, objectives, and other accessories sold separately.