VWR® Talon® Regular Clamp Holder
Proveedor: VWR
VWR® Talon®
80862553 80862716
89202-610EA 1983 MXN
89202-610 21572-501
VWR® Talon® Regular Clamp Holder
Abrazaderas Adaptadores en cruz
Features a streamlined design for maximum contamination resistance and easy cleaning.
  • Use wherever clamping at 90° is required
  • Available in nickel-plated zinc or electropolished stainless steel finish
  • Strong and durable
Regular clamp holders are used to hold extension clamps to lab frames where 90° positioning is required.