VWR® Odor Eliminators
Proveedor: VWR
VWR®
04393-65 04393-61 04393-68 04393-62
80080-890CS 7879.18 MXN
80080-890 80080-896 80080-898 80080-888
VWR® Odor Eliminators
Desodorantes
These sterilizer deodorizers chemically absorb odor-causing molecules, removing unwanted odors from your facility rather than masking them.
  • Chemically absorbent deodorizers
Simply place one capsule in your autoclave or incubator with each load. The heat and moisture will dissolve the capsule and release a clean fresh scent.