Nalgene® Desiccators, 250mm, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
5309-0250 5310-0250
24987-026CS 16746.21 MXN
24987-026 24987-004
Nalgene® Desiccators, 250mm, Thermo Scientific
Desecadores Desecadores de plástico
Transparent polycarbonate cover and polypropylene base will not shatter or implode at full vacuum when used according to instructions packed with product
Desiccator 24987-004 has a Teflon® resin stopcock plug that provides convenient vacuum connection on body, making cover easy to handle. Straight-bore design eliminates stopcock leakage. Use with 1/4" I.D. vacuum tubing.
Body is impervious to all the usual desiccants except concentrated acids. No gasket necessary, and no need to draw a vacuum to make cover airtight; only grease is needed. Unique recessed-rim cover keeps grease off bench when cover is set down.
Información para pedidos: Desiccators take any standard 230mm desiccator plate, such as 25038-003. Desiccator 24987-004 has a stopcock plug for vacuum connection and is not autoclavable (for room temperature use only). Desiccator 24987-026 does not include a stopcock and is autoclavable.