Instrument Trays, Stainless Steel, Polar ware
Proveedor: Polar Ware
E1650 E1654 1202
62687-070CS 8015.33 MXN
62687-070 62687-049 62687-069
Instrument Trays, Stainless Steel, Polar ware
Bandejas/Platos/Ollas Bandejas/Placas de instrumental
For sterilization of instruments, syringes, and more
Made of stainless steel for easy cleaning and quick sterilization. Stain-proof and long lasting — won’t chip, break, or corrode. May be ordered with or without solid flat covers. Same cover is used for trays 62687-069 and -070.