DRIERITE® Gas Drying Unit, W.A. Hammond Drierite Co
26800
26668-007CS 14692.33 MXN
26668-007
DRIERITE® Gas Drying Unit, W.A. Hammond Drierite Co
Refillable polycarbonate column filled with Indicating DRIERITE®.
  • Dries air and gases saturated with moisture to a dew point of approximately –73°C (–100°F)
  • Desiccant will turn from blue to pink as it is exhausted
  • Column measures 2.5” dia. x 11.5”H, and features two hose barb connections
  • Unit holds 0.6kg (1.25lbs.) of 8 mesh indicating DRIERITE® desiccant (included)
  • Flow rates up to 3.3 lpm
  • Can be used under vacuum
Drierite can be regenerated by removing from column and heating granules on a tray in a single layer at 210–220°C for 1 hour. Cool in a sealed container before re-packing drying column. This procedure is very specific.