VWR® Desiccator Plates
Proveedor: VWR
VWR®
J-162 J-161 J-160 9180003210 J-159
89038-074CS 4311.57 MXN
89038-074 89038-072 76376-702 89038-070 89038-068
VWR® Desiccator Plates
Desecadores Placas de desecadores
Chemical-porcelain dessicator plates are perforated with numerous small holes and a large center hole.
  • Autoclavable
  • Plates without feet
Plates are also glazed on one side and highly resistant to chemicals. For use in temperatures up to 1150 °C (2102 °F).