VWR® Sterile Cell Spreaders, PP
Proveedor: VWR
VWR®
24408-21
89042-021PK 391.81 MXN
89042-021
VWR® Sterile Cell Spreaders, PP
Esparcidores Esparcidores de células
Disposable, L-shaped cell spreaders are ideal for spreading bacterial cultures evenly.
  • L-shape allows application on the entire Petri dish or plate surface
  • Smooth spreading surface with slight upward return reduces the risk of damaging agar samples
Spreaders are presterilized, eliminating the need for flaming or autoclaving.
Envase: Packaged in resealable bags of 10.