VWR® Reusable Inoculating Loops and Needles
Proveedor: Avantor
VWR®
24408-29 24408-30 24408-34 24406-80 24408-24 24408-33 24408-32 24408-31 24408-27 24408-25 24408-23 24406-79 24408-22 24408-35 24408-26
80094-480PK 1371.87 MXN
80094-480 80094-492 80094-484 80094-496 80094-482 80094-494 80094-488 80094-510 80094-478 80094-486 80094-498 80094-476 80094-504 80094-507 80094-508
VWR® Reusable Inoculating Loops and Needles
Esparcidores Asas de siembra
Inoculating loops and needles are constructed of high-quality, economic nichrome wire that cools rapidly after sterilization.
  • Reusable
  • Lightweight
  • Aluminum handles are available with loops and needles
  • Nickel-plated brass construction of loop holders
Loops slide along agar surfaces without cutting or tearing
Calibrated loops are supplied with certificates based on ASM/APHA Evans blue dye test protocols.
Loops are available with or without lightweight aluminum handles, which are insulated for hand protection. Reusable needles feature lightweight, insulated aluminum handles and are ideal for selecting, inoculating, or transferring microbiological samples. Loop holders are constructed of insulated nickel-plated brass.