VWR® Inoculating Loops and Needles, Sterile
Proveedor: Avantor
VWR®
24406-58 24406-59 24406-63 24406-57 24406-64 24406-62 24406-56 24406-60 24406-61
12000-806CS 1588.44 MXN
12000-806 12000-808 89126-870 89126-872 12000-810 12000-812 89126-868 12000-814 12000-816
VWR® Inoculating Loops and Needles, Sterile
Esparcidores Asas de siembra
High-impact polystyrene loops and needles are smooth and flexible to facilitate uniform and smooth streaking without damaging the gel surface.
  • Polystyrene fabrication
  • Smooth and flexible
  • Sterile
Needles are straight and suitable for removal of single colony specimens. Packed sterile in safe, tamperproof, zip-seal resealable bags. Disposable inoculating loops and needles do not need flaming, eliminating the risk of infection due to aerosol formation of pathogenic substances as well as cross-contamination due to improper sterilization. Loops and needles can be used under hoods without danger. Color-coded for easy size identification. Total length: 22.7 cm (87/8").