VWR® Signature™ Inoculating Loops and Needles, Sterile
Proveedor: VWR
VWR® Signature™
175CS50T.VWR 177CS50T.VWR 179CS50T.VWR 176CS50T.VWR 178CS50T.VWR
30002-096CS 3753.41 MXN
30002-096 30002-098 30002-092 30002-094 30002-090
VWR® Signature™ Inoculating Loops and Needles, Sterile
Esparcidores Asas de siembra
Calibrated disposable polystyrene inoculation loops and needles.
  • Disposable polystyrene fabrication
  • Color-coded for easy size identification
  • Available in soft or rigid plastic
  • Ultra-smooth surface for easy streaking
Loops are available in soft or rigid plastic and feature an ultra smooth surface for easy streaking. Loops and needles are color-coded for easy size identification.
Envase: Packaged in anti-roll tubes with push-on resealable cap, 50 loops/needles per tube, 20 tubes per case. Length: 21 cm (81/4").