Vacuum Trap with Serrated Connections, Chemglass
Proveedor: Chemglass
CG-4515-20 CG-4515-03 CG-4515-02 CG-4515-01 CG-4515-04
80072-548EA 3534.36 MXN
80072-578 80072-548 80072-552 80072-554 80072-550
Vacuum Trap with Serrated Connections, Chemglass
Trampas de vacío
Two-piece construction for easy removal of condensate
Inlet and outlet are 180° apart for installation into a variety of vacuum systems. Stopper has a [ST] inner joint and body has an outer joint. Body length is measured below the joint. Serrated hose connections have an O.D. of 13mm at the smallest serration and 16mm at the largest serration.