WHEATON® Serum Vials, Borosilicate Glass, DWK Life Sciences
Proveedor: DWK Life Sciences
223686 223695 223696 223693 223684 223683 223685 223687
16171-287CS 2198.2 MXN
16171-285 16171-286 16171-287 16171-298 16171-288 16171-293 16171-284 16171-291
WHEATON® Serum Vials, Borosilicate Glass, DWK Life Sciences
Viales
Tubular design provides excellent clarity and dimensional consistency from vial to vial.
  • Autoclavable
Clear vials manufactured from low extractable borosilicate glass.Amber glass protect light-sensitive samples.
Specially designed bottom radius adds strength for lyophilization applications.
Certificaciones: Amber glass conforms to USP Type I requirements.Borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Envase: Vials are packaged in shrink-wrapped modules reduce particulate contamination.