Puritan® Standard Cotton Tipped Applicators, Wood Handle, Sterile, Puritan Medical Products
Proveedor: Puritan Medical Products
Puritan®
25-8062WC 25-8061WC 25-80610WC 25-80610WC 25-8062WC 25-806-1WC
10805-154CS 3354.12 MXN
89133-752 CA89133-752 CA10805-156 CA10805-154 10805-156 10805-154
Puritan® Standard Cotton Tipped Applicators, Wood Handle, Sterile, Puritan Medical Products
Hisopos Hisopos para muestras
This absorbent, cotton tipped applicator is produced with high-quality USP grade cotton which is carded and coiled onsite to maintain consistency
Designed for general-purpose use, specimen collection, cell collection for DNA testing, site preparation, cosmetic applications, medication applicator, and for wound dressing and cleaning. Made in the USA.
It is bonded securely to a rigid white birch handle by an aqueous based adhesive.
Certificaciones: CE marked.