SP Bel-Art LABMAT™ Bench Liner, Bel-Art Products, a part of SP
Proveedor: Bel-Art
LABMAT™
F24675-0000 F24675-0000_SE F24675-0020 F24675-1000
54110-300CS 10867.35 MXN
54110-300 470145-292 13196-048 47727-664
SP Bel-Art LABMAT™ Bench Liner, Bel-Art Products, a part of SP
Protectores de superficies Protectores de sobremesa
Disposable Labmat™ Bench Liner speeds clean-ups and safeguards work surfaces from stains, spills, and wear.
  • Disposable
  • Supplied in dispenser carton
  • Absorbent upper surface and flexible plastic non-skid backing
  • Roll length: 15.2 m (50')
The highly absorbent upper surface traps up to 420ml/m2 of water, dyes, corrosive chemicals, or radioactive solutions. The flexible polyethylene film backing is non-skid and prevents soak-through of most solutions. Simply roll up and discard after contamination. Labmat™ Liner is available in 50.8cm (20”) wide by 15.2m (50ft.) rolls or 50.8 x 61cm (20 x 24” ) pre-cut sheets. Rolls are easily cut with scissors while roll is retained in dispenser.
Envase: 50 ft. roll in dispenser carton (6 cartons per case) or box of 100 sheets.