VWR® Critical Swab® Swab, Medium Foam Head
TX815V
89022-990CS 16039.75 MXN
89022-990
VWR® Critical Swab® Swab, Medium Foam Head
Swab features 100ppi open cell polyurethane foam with a long, 100% virgin polypropylene handle.
  • Polyurethane foam head
  • Polypropylene handle
  • No adhesive
Swab head is thermally bonded to the handle using no adhesive.